Aluminium chloride, anhydrous ≥99%

Supplier: BeanTown Chemical

Synonyms: Aluminum chloride

218275-5KG 218275-1KG 218275-250G
BT218275-5KGEA 364.15 USD
BT218275-5KG BT218275-1KG BT218275-250G
Aluminium chloride, anhydrous ≥99%
Aluminium chloride

  • RTECS: BD0525000
  • Moisture Sensitive
  • Granular

Formula: AlCl₃
MW: 133.34 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR