Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicon ≥99.999% (trace metals basis), crystalline powder -100+435 mesh

Supplier: BeanTown Chemical


218225-10G 218225-50G 218225-250G
BT218225-10GEA 64.42 USD InStock
BT218225-10G BT218225-50G BT218225-250G
Silicon ≥99.999% (trace metals basis), crystalline powder -100+435 mesh
Silicon
CAS: 7440-21-3; EC No: 231-130-8; MDL No: MFCD00085311; RTECS: VW0400000
UN No: UN1346; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Si; MW: 28.09
Melting Point: 1410°; Boiling Point: 2355°
Density (g/mL): 2.33


Formula: Si
MW: 28,09 g/mol
Storage Temperature: ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty