Ytterbium(III) nitrate hydrate ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Ytterbium trinitrate hydrate
218215-50G
218215-10G
BT218215-50GEA
158.68
USD
BT218215-50G
BT218215-10G
Ytterbium(III) nitrate hydrate ≥99.9% (trace rare earth metals basis)
Ytterbium(III) nitrate hydrate
- Hygroscopic
- Crystalline
Formula:
Yb(NO₃)₃·xH₂O MW: 359.05 g/mol (anhy) |
MDL Number:
MFCD00149883 EINECS: 237-384-6 ADR: 5.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...