(±)-Methyl-2,3-dichloropropionate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyl 2,3-dichloropropanoate
218105-25G
218105-500G
218105-100G
BT218105-25GEA
45.7
USD
BT218105-25G
BT218105-500G
BT218105-100G
(±)-Methyl-2,3-dichloropropionate ≥98%
(±)-Methyl 2,3-dichloropropionate
- Refractive index: 1.453
- Liquid
Formula:
C₄H₆Cl₂O₂ MW: 157 g/mol Boiling Pt: 52…55 °C/2 mmHg Density: 1.325 g/mL Flash Pt: 63 °C (145 °F) |
MDL Number:
MFCD00000944 CAS Number: 3674-09-7 EINECS: 222-940-2 UN: 3265 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...