Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Praseodymium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical


218075-50X50MM 218075-25X25MM
BT218075-50X50EA 494.42 USD InStock
BT218075-50X50 BT218075-25X25
Praseodymium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Praseodymium
CAS: 7440-10-0; EC No: 231-120-3; MDL No: MFCD00011174
UN No: UN3178; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: Pr; MW: 140.91
Melting Point: 931°; Boiling Point: 3520°
Density (g/mL): 6.71


Formula: Pr
MW: 140,91 g/mol
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty