Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,6-Diaminohexane ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,6-HexanediamineHexamethylenediamine

217995-100G 217995-2.5KG 217995-500G
BT217995-100GEA 17.87 USD InStock
BT217995-100G BT217995-2.5KG BT217995-500G
1,6-Diaminohexane ≥98%
1,6-Diaminohexane
CAS: 124-09-4; EC No: 204-679-6; MDL No: MFCD00008243; RTECS: MO1180000
UN No: UN2280; Haz Class: 8; Packing Group: III
Solid; Linear Formula: NH2(CH2)6NH2; Molecular Formula: C6H16N2; MW: 116.20
Melting Point: 42-45°; Boiling Point: 204-205°; Flash point: 80°C (176°F)
Density (g/mL): 0.89; Refractive Index: 1.439
Air Sensitive , Hygroscopic


Formula: C₆H₁₆N₂
MW: 116,21 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 39 to 42 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: ambient
MDL Number: MFCD00008243
CAS Number: 124-09-4
EINECS: 204-679-6
UN: 2280
ADR: 8,III
Merck Index: 13,04713
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty