Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethylzinc ≥96%

Supplier: BeanTown Chemical


217810-25G
BT217810-25GEA 1080.09 USD InStock
BT217810-25G
Dimethylzinc ≥96%
Dimethylzinc
CAS: 544-97-8; EC No: 208-884-1; MDL No: MFCD00014854
UN No: UN3394; Haz Class: 4.2 (4.3); Packing Group: I
Liquid, packaged in a non-returnable cylinder; Linear Formula: (CH3)2Zn; MW: 95.44
Boiling Point: 46°; Flash point: -18°C (0°F)
Density (g/mL): 1.386
Air Sensitive, Moisture Sensitive


Formula: C₂H₆Zn
MW: 95,478 g/mol
MDL Number: MFCD00014854
CAS Number: 544-97-8
EINECS: 208-884-1
Order Now
Specification Test Results
Assay 96%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty