Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc bromide, anhydrous ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Zinc (II) bromide

Ampuled under argon

217795-100G 217795-5G 217795-25G
BT217795-100GEA 774.87 USD InStock
BT217795-100G BT217795-5G BT217795-25G
Zinc bromide, anhydrous ≥99.999% (trace metals basis)
Zinc bromide
CAS: 7699-45-8; EC No: 231-718-4; MDL No: MFCD00011294; RTECS: ZH1150000
UN No: UN3260; Haz Class: 8; Packing Group: II
Beads; Linear Formula: ZnBr2; MW: 225.19
Melting Point: 394°; Boiling Point: 697°
Density (g/mL): 4.2; Refractive Index: 1.545
Moisture sensitive


Formula: Br₂Zn
MW: 225,2 g/mol
Boiling Pt: 670 to 680 °C (1013 hPa)
Melting Pt: 394 °C
Density: 4.22 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 13,10182
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty