Methylmercury chloride 1000 ppm by AA in water

Danger

217755-5ML 217755-25ML
BT217755-25MLEA 270.03 USD
BT217755-25ML BT217755-5ML
Methylmercury chloride 1000 ppm by AA in water
Methylmercury chloride
Formula: CH₃ClHg
MW: 251.08 g/mol
Melting Pt: 170 …173 °C
MDL Number: MFCD00013592
CAS Number: 115-09-3
UN: 2024
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR