Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentanol ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: n-Amyl alcohol1-Pentyl alcoholAmyl alcohol1-Hydroxy pentaneButyl carbinol

217605-250ML 217605-1L 217605-4L
BT217605-250MLEA 23.38 USD InStock
BT217605-250ML BT217605-1L BT217605-4L
1-Pentanol ≥99.0% ACS
1-Pentanol
CAS: 71-41-0; EC No: 200-752-1; MDL No: MFCD00002977; RTECS: SB9800000
UN No: UN1105; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)4OH; Molecular Formula: C5H12O ; MW: 88.15
Melting Point: -78°; Boiling Point: 136-138°; Flash point: 49°C (120°F)
Density (g/mL): 0.811; Refractive Index: 1.409


Formula: H₃C(CH₂)₃CH₂OH
MW: 88,15 g/mol
Boiling Pt: 137.9 °C (1013 hPa)
Melting Pt: -79 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3,III
Merck Index: 13,07195
https://us.vwr.com/stibo/web/std.lang.all/15/15/731515.jpg
Order Now
Specification Test Results
Assay 99.0% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty