Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium fluoride, anhydrous ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Aluminum fluoride

217570-10G 217570-50G 217570-250G
BT217570-10GEA 82.34 USD InStock
BT217570-10G BT217570-50G BT217570-250G
Aluminium fluoride, anhydrous ≥99.99% (trace metals basis)
Aluminium fluoride
CAS: 7784-18-1; EC No: 232-051-1; MDL No: MFCD00003426
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: AlF3; MW: 83.98
Melting Point: 1291°
Density (g/mL): 3.1
Hygroscopic


Formula: AlF₃
MW: 83,98 g/mol
Melting Pt: 1040 °C
Density: 2.88 g/cm³ (20 °C)
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8,II
Merck Index: 12,00351
Order Now
Specification Test Results
Assay 99.99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty