Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Praseodymium(III) nitrate hydrate ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical

Synonyms: Praseodymium nitrate hydrateNitricacid, praseodymium(3+) salt (3:1)

217540-50G 217540-10G 217540-250G
BT217540-50GEA 56.89 USD InStock
BT217540-50G BT217540-10G BT217540-250G
Praseodymium(III) nitrate hydrate ≥99.9% (trace rare earth metals basis)
Praseodymium(III) nitrate hydrate
EC No: 233-796-5; MDL No: MFCD00011175
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Pr(NO3)3·xH2O; MW: 326.92 (anhydrous)
Hygroscopic


Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty