Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethylamine 40% (w/w) in water

Supplier: BeanTown Chemical


217530-250G 217530-1KG
BT217530-250GEA 18.69 USD InStock
BT217530-250G BT217530-1KG
Dimethylamine 40% (w/w) in water
Dimethylamine
CAS: 124-40-3; EC No: 204-697-4; MDL No: MFCD00008288
UN No: UN1160; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: (CH3)2NH; MW: 45.08
Flash point: 16°C (61°F)
Density (g/mL): 0.89; Refractive Index: 1.370


Formula: C₂H₇N
MW: 45.08 g/mol
Boiling Pt: 6.9 °C (1013 hPa)
Melting Pt: -92.8 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: -55 °C
Storage Temperature: ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3,II
Merck Index: 12,03278
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty