Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4,4'-Trichlorocarbanilide ≥99%

Supplier: BeanTown Chemical

Synonyms: Triclocarban3,4,4'-Trichlorodiphenylurea1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea

217510-100G
BT217510-100GEA 51.85 USD InStock
BT217510-100G
3,4,4'-Trichlorocarbanilide ≥99%
3,4,4'-Trichlorocarbanilide
CAS: 101-20-2; MDL No: MFCD00013254
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Linear Formula: Cl2C6H3NHCONHC6H4Cl; Molecular Formula: C13H9Cl3N2O; MW: 315.58
Melting Point: 254-256°; Flash point: 150°C (302°F)
Density (g/mL): 1.53


Formula: C₁₃H₉Cl₃N₂O
MW: 315,58 g/mol
MDL Number: MFCD00013254
CAS Number: 101-20-2
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty