Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrophenyl selenocyanate ≥97%

Supplier: BeanTown Chemical


217430-1G 217430-5G
BT217430-1GEA 54.12 USD InStock
BT217430-1G BT217430-5G
2-Nitrophenyl selenocyanate ≥97%
2-Nitrophenyl selenocyanate
CAS: 51694-22-5; EC No: 257-350-4; MDL No: MFCD00043146
UN No: UN3283; Haz Class: 6.1; Packing Group: II
Crystalline/Solid; Molecular Formula: C7H4N2O2Se; MW: 227.08
Melting Point: 140-142°


Formula: C₇H₄N₂O₂Se
MW: 227,092 g/mol
MDL Number: MFCD00043146
CAS Number: 51694-22-5
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty