2-Chlorobutane ≥98%

Supplier: BeanTown Chemical

217415-250ML 217415-50ML
BT217415-50MLEA 24.92 USD
BT217415-50ML BT217415-250ML
2-Chlorobutane ≥98%
2-Chlorobutane

  • RTECS: EJ6475000
  • Refractive index: 1.396
  • Liquid

Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68…70 °C
Melting Pt: –140 °C
Density: 0.873 g/mL
Flash Pt: –15 °C (5 °F)
MDL Number: MFCD00000871
CAS Number: 78-86-4
EINECS: 201-151-7
UN: 1127
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR