Pyrrole ≥98%

Supplier: BeanTown Chemical

Synonyms: 1H-Pyrrole

217365-1KG 217365-50G 217365-250G
BT217365-1KGEA 200.32 USD
BT217365-1KG BT217365-250G BT217365-50G
Pyrrole ≥98%
Pyrrole

  • RTECS: UX9275000
  • Refractive index: 1.508
  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Liquid

Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 131 °C
Melting Pt: –23 °C
Density: 0.967 g/mL
Flash Pt: 36 °C (97 °F)
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR