Sodium periodate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Sodium metaperiodate
217355-100G
217355-500G
217355-10G
BT217355-100GEA
67.91
USD
BT217355-100G
BT217355-500G
BT217355-10G
Sodium periodate ≥98%
Sodium periodate
- RTECS: SD4550000
- Hygroscopic, Light Sensitive
- Crystalline/Powder
Formula:
NaIO₄ MW: 213.89 g/mol Melting Pt: 300 °C Density: 3.86 g/mL |
MDL Number:
MFCD00003534 CAS Number: 7790-28-5 EINECS: 232-197-6 UN: 1479 ADR: 5.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...