Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Europium(II) fluoride ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical


217290-500MG 217290-10G 217290-2G
BT217290-500MGEA 130.65 USD InStock
BT217290-500MG BT217290-10G BT217290-2G
Europium(II) fluoride ≥99.9% (trace metals basis)
Europium (II) fluoride
CAS: 14077-39-5; MDL No: MFCD01866333
UN No: UN3288; Haz Class: 6.1; Packing Group: III
-200 Mesh Powder; Linear Formula: EuF2; MW: 189.96
Hygroscopic


Formula: EuF₂
MW: 189,96 g/mol
MDL Number: MFCD01866333
CAS Number: 14077-39-5
EINECS: 000-000-0
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty