Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc chloride, anhydrous ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Danger
Synonyms: Zinc (II) chlorideButter of zinc

Ampuled under argon

217250-25G 217250-5G 217250-100G
BT217250-25GEA 201.6 USD InStock
BT217250-25G BT217250-5G BT217250-100G
Zinc chloride, anhydrous ≥99.999% (trace metals basis)
Zinc chloride
CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295; RTECS: ZH1400000
UN No: UN2331; Haz Class: 8; Packing Group: III
Beads; Linear Formula: ZnCl2; MW: 136.29
Melting Point: 293°; Boiling Point: 732°
Density (g/mL): 2.907
Hygroscopic


Formula: ZnCl₂
MW: 136,3 g/mol
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty