Triethyltin chloride
217135-1G
217135-25G
BT217135-1GEA
17.85
USD
BT217135-1G
BT217135-25G
Triethyltin chloride
Triethyltin chloride
- Moisture Sensitive
- Liquid
Formula:
C₆H₁₅ClSn MW: 241.35 g/mol Boiling Pt: 260 °C Density: 1.429 g/mL |
MDL Number:
MFCD00040414 CAS Number: 994-31-0 EINECS: 213-616-1 UN: 2788 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...