Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) metaborate monohydrate

Supplier: BeanTown Chemical


217055-25G 217055-100G
BT217055-25GEA 85.27 USD InStock
BT217055-25G BT217055-100G
Lead(II) metaborate monohydrate
Lead(II) metaborate monohydrate
CAS: 10214-39-8; EC No: 238-762-3; MDL No: MFCD00049504
UN No: UN2291; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: PbB2O4; MW: 310.81


Formula: B₂O₄Pb·H₂O
MW: 310,81 (292,81 anhy)
MDL Number: MFCD00049504
CAS Number: 10214-39-8
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty