Phosphorus(V) chloride ≥99.995% (trace metals basis)

216945-100G 216945-25G
BT216945-25GEA 110.36 USD
BT216945-25G BT216945-100G
Phosphorus(V) chloride ≥99.995% (trace metals basis)
Phosphorus (V) chloride

  • RTECS: TB6125000
  • Moisture Sensitive
  • Powder

Formula: Cl₅P
MW: 208.24 g/mol
Melting Pt: 160 °C (subl.)
Density: 2.114 g/mL
MDL Number: MFCD00011439
CAS Number: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8,II

Order Now

Specification Test Results

Assay 99.995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR