Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) chloride, anhydrous ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Cobalt dichlorideCobaltous chloride

Ampuled under argon

216900-5G 216900-25G 216900-1G
BT216900-5GEA 172.31 USD InStock
BT216900-5G BT216900-25G BT216900-1G
Cobalt(II) chloride, anhydrous ≥99.999% (trace metals basis)
Cobalt (II) chloride
CAS: 7646-79-9; EC No: 231-589-4; MDL No: MFCD00010938; RTECS: GF9800000
UN No: UN3077; Haz Class: 9; Packing Group: III
-10 Mesh Beads; Linear Formula: CoCl2; MW: 129.84
Melting Point: 724°
Hygroscopic


Formula: Cl₂Co
MW: 129,84 g/mol
Boiling Pt: 1049 °C (1013 hPa)
Melting Pt: 735 °C
Density: 1.6795 g/cm³ (20 °C)
Storage Temperature: 2 - 8°C
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 12,02498
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty