Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodomethane ≥99% stabilized

Supplier: BeanTown Chemical

Synonyms: Methyl iodide

216890-250G 216890-1KG 216890-50G
BT216890-250GEA 66.88 USD InStock
BT216890-250G BT216890-1KG BT216890-50G
Iodomethane ≥99% stabilized
Iodomethane
CAS: 74-88-4; EC No: 200-819-5; MDL No: MFCD00001073; RTECS: PA9450000
UN No: UN2644; Haz Class: 6.1; Packing Group: I
Liquid; Molecular Formula: CH3I; MW: 141.94
Melting Point: -64°; Boiling Point: 41-43°
Density (g/mL): 2.28; Refractive Index: 1.531
Light Sensitive


Formula: CH₃I
MW: 141,94 g/mol
Boiling Pt: 42.4 °C (1013 hPa)
Melting Pt: -66 °C
Density: 2.279 g/cm³ (20 °C)
Storage Temperature: 2 - 8°C
MDL Number: MFCD00001073
CAS Number: 74-88-4
EINECS: 200-819-5
UN: 2644
ADR: 6.1,I
Merck Index: 12,06161
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty