(S)-(-)-3-Methoxy-α-methylbenzylamine ≥99%, ee 99% ee
Synonyms:
(S)-(-)-1-(3-Methoxyphenyl)ethylamine
216800-5G
216800-25G
216800-1G
BT216800-1GEA
47.14
USD
BT216800-1G
BT216800-5G
BT216800-25G
(S)-(-)-3-Methoxy-α-methylbenzylamine ≥99%, ee 99% ee
(S)-(-)-3-Methoxy-α-methylbenzylamine
- Air Sensitive
- Liquid
Formula:
C₉H₁₃NO Density: 1.023 g/mL Flash Pt: 114 °C (237 °F) |
MDL Number:
MFCD00671656 CAS Number: 82796-69-8 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...