Methyl oxalyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
Methyl chloroglyoxylate
, Methyl 2-chloro-2-oxoacetate
216780-25G
216780-5G
216780-100G
BT216780-25GEA
120.16
USD
BT216780-25G
BT216780-5G
BT216780-100G
Methyl oxalyl chloride ≥97%
Methyl oxalyl chloride
- Refractive index: 1.419
- Moisture Sensitive
- Liquid
Formula:
C₃H₃ClO₃ MW: 122.51 g/mol Boiling Pt: 118…120 °C Density: 1.332 g/mL Flash Pt: 46 °C (114 °F) |
MDL Number:
MFCD00000705 CAS Number: 5781-53-3 EINECS: 227-307-4 UN: 2920 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...