Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Arsenic ≥99.99999% (trace metals basis), polycrystalline lumps 2-8 mm

Supplier: BeanTown Chemical


Ampuled under argon

216670-10G 216670-25G
BT216670-10GEA 109.03 USD InStock
BT216670-10G BT216670-25G
Arsenic ≥99.99999% (trace metals basis), polycrystalline lumps 2-8 mm
Arsenic
CAS: 7440-38-2; EC No: 213-148-6; MDL No: MFCD00085309; RTECS: CG0525000
UN No: UN1558; Haz Class: 6.1; Packing Group: II
Solid; Molecular Formula: As ; MW: 74.92
Melting Point: 817°; Boiling Point: 613° (sublimes)
Density (g/mL): 5.727


Formula: As
MW: 74.92 g/mol
Melting Pt: 817 °C
Density: 5.727 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 231-148-6
UN: 1558
ADR: 6.1,II
Merck Index: 12,00832
Order Now
Specification Test Results
Assay 99.99999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty