Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromium(VI) oxide ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Chromic anhydrideChromium trioxide

216635-100G 216635-500G
BT216635-100GEA 51.03 USD InStock
BT216635-100G BT216635-500G
Chromium(VI) oxide ≥98.0% ACS
Chromium (VI) oxide
CAS: 1333-82-0; EC No: 215-607-8; MDL No: MFCD00010952; RTECS: GB6650000
UN No: UN1463; Haz Class: 5.1 (6.1,8); Packing Group: II
Crystalline; Molecular Formula: CrO3; MW: 99.99
Melting Point: 196° (decomposes)
Density (g/mL): 2.7
Hygroscopic


Formula: CrO₃
MW: 99,99 g/mol
Melting Pt: 195 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00010952
CAS Number: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1,II
REACH: 01-2119458868-17
Merck Index: 13,02256
Order Now
Specification Test Results
Assay 98.0% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty