Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylammonium hydroxide 25% (w/w) in methanol

Supplier: BeanTown Chemical


216580-100G 216580-500G
BT216580-100GEA 33.27 USD InStock
BT216580-100G BT216580-500G
Tetramethylammonium hydroxide 25% (w/w) in methanol
Tetramethylammonium hydroxide
CAS: 75-59-2; EC No: 200-882-9; MDL No: MFCD00008280
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Linear Formula: (CH3)4NOH ; Molecular Formula: C4H13NO; MW: 91.15
Flash point: 12°C (54°F)
Density (g/mL): 0.866
Air Sensitive , Hygroscopic


Formula: C₄H₁₃NO
MW: 91,15 g/mol
Melting Pt: 67 to 70 °C
Density: 1.016 g/cm³ (25 °C)
Storage Temperature: ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 2924
ADR: 3,II
Merck Index: 14,09224
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty