Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium acetylacetonate hydrate

Supplier: BeanTown Chemical


216305-25G 216305-100G
BT216305-25GEA 62.98 USD InStock
BT216305-25G BT216305-100G
Barium acetylacetonate hydrate
Barium acetylacetonate hydrate
CAS: 12084-29-6; EC No: 235-151-3; MDL No: MFCD00013484
UN No: UN1564; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C10H14BaO4·xH2O; MW: 335.54 (anhydrous)


Formula: C₁₀H₁₄BaO₄·xH₂O
MW: 335,545 g/mol (anhydrous)
MDL Number: MFCD00013484
CAS Number: 12084-29-6
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty