Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl-3-aminoazetidine-1-carboxylate ≥94%

Supplier: BeanTown Chemical

Synonyms: 1-BOC-3-aminoazetidine3-Amino-1-Boc-azetidine

216275-1G 216275-250MG
BT216275-1GEA 432.49 USD InStock
BT216275-1G BT216275-250MG
tert-Butyl-3-aminoazetidine-1-carboxylate ≥94%
tert-Butyl-3-aminoazetidine-1-carboxylate
CAS: 193269-78-2; MDL No: MFCD01861753
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C8H16N2O2; MW: 172.23


Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
MDL Number: MFCD01861753
CAS Number: 193269-78-2
Order Now
Specification Test Results
Assay 94%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty