Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium ≥99% (trace metals basis), rods Ø 22mm (max. 0.8% Sr)

Supplier: BeanTown Chemical


216270-100G 216270-500G
BT216270-100GEA 153.17 USD InStock
BT216270-100G BT216270-500G
Barium ≥99% (trace metals basis), rods Ø 22mm (max. 0.8% Sr)
Barium
CAS: 7440-39-3; EC No: 231-149-1; MDL No: MFCD00134031; RTECS: CQ8370000
UN No: UN1400; Haz Class: 4.3; Packing Group: II
Solid; Molecular Formula: Ba; MW: 137.33
Melting Point: 725°; Boiling Point: 1640°
Density (g/mL): 3.6


Formula: Ba
MW: 137,33 g/mol
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty