Propagyl bromide ≥80% (w/w) in toluene stabilized

Supplier: BeanTown Chemical

Synonyms: 1-Brom-2-propin3-Bromopropyne

216215-1L 216215-250ML 216215-50ML
BT216215-1LEA 683.46 USD
BT216215-1L BT216215-250ML BT216215-50ML
Propagyl bromide ≥80% (w/w) in toluene stabilized
Propagyl bromide

  • RTECS: UK4375000
  • Refractive index: 1.494
  • Light Sensitive
  • Liquid

Formula: C₃H₃Br
Boiling Pt: 88…90 °C
Melting Pt: –61 °C
Density: 1.38 g/mL
Flash Pt: 3 °C (37 °F)
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR