Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) chloride ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Antimony trichloride

Ampuled under argon

216000-100G 216000-25G 216000-500G
BT216000-100GEA 418.96 USD InStock
BT216000-100G BT216000-25G BT216000-500G
Antimony(III) chloride ≥99.999% (trace metals basis)
Antimony (III) chloride
CAS: 10025-91-9; EC No: 233-047-2; MDL No: MFCD00011212; RTECS: CC4900000
UN No: UN1733; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: SbCl3; MW: 228.12
Melting Point: 73.4°
Moisture Sensitive


Formula: SbCl₃
MW: 228,12 g/mol
Boiling Pt: 223.5 °C (1013 hPa)
Melting Pt: 73 °C
Density: 3.14 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty