Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium dihydrogen phosphate 10% in nitric acid 2% Matrix Modifier Solution

Supplier: BeanTown Chemical

Synonyms: Ammonium phosphate, monobasicAmmonium Phosphate Monobasic

215895-100ML
BT215895-100MLEA 131.29 USD InStock
BT215895-100ML
Ammonium dihydrogen phosphate 10% in nitric acid 2% Matrix Modifier Solution
Ammonium dihydrogen phosphate
EC No: 231-714-2; MDL No: MFCD00003396
UN No: UN3264; Haz Class: 8; Packing Group: III


Formula: NH₄H₂PO₄
MW: 115,03 g/mol
Melting Pt: 190 °C
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty