Erbium(III) perchlorate hexahydrate 50% (w/w) in water

Supplier: BeanTown Chemical

215850-C10G 215850-C2G 215850-C50G
BT215850-C50GEA 569.2 USD
BT215850-C50G BT215850-C2G BT215850-C10G
Erbium(III) perchlorate hexahydrate 50% (w/w) in water
Erbium (III) perchlorate hexahydrate

  • Liquid

Formula: Cl₃ErO₁₂·6H₂O
MW: 573.7 g/mol
MDL Number: MFCD00150232
CAS Number: 14692-15-0
EINECS: 237-840-4
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR