Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylammonium heptadecafluorooctane-1-sulfonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Heptadecafluorooctanesulphonic acid tetraethylammonium saltTetraethylammonium perfluorooctane-1-sulfonatePerfluoro-1-octanesulfonic acid tetraethylammonium saltTetraethylammonium perfluorooctanesulphonateTetraethylammonium perfluorooctane-1-sulphonateHeptadecafluoro-1-octanesulfonic acid tetraethylammonium saltHeptadecafluoro-1-octanesulphonic acid tetraethylammonium saltPerfluoro-1-octanesulphonic acid tetraethylammonium salt

215805-5G
BT215805-5GEA 43.52 USD InStock
BT215805-5G
Tetraethylammonium heptadecafluorooctane-1-sulfonate ≥98%
Tetraethylammonium heptadecafluorooctane-1-sulphonate
CAS: 56773-42-3; EC No: 260-375-3; MDL No: MFCD00066497; RTECS: KH3154250
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: CF3(CF2)7SO3N(CH2CH3)4; Molecular Formula: C16H20F17NO3S; MW: 629.37
Melting Point: 184-190°
Hygroscopic


Formula: C₁₆H₂₀F₁₇NO₃S
MW: 629,38 g/mol
CAS Number: 56773-42-3
EINECS: 260-375-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty