Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄) (w/w)
Supplier: BeanTown Chemical
|
|
215685-50ML
215685-250ML
BT215685-250MLEA
238.12
USD
BT215685-250ML
BT215685-50ML
Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄) (w/w)
Tetrafluoroboric acid-diethyl ether complex (50-55% tetrafluoroboric acid in diethyl ether)
- Liquid
Formula:
C₄H₁₁BF₄O MW: 161.93 g/mol Density: 1.19 g/mL Flash Pt: 36 °C (97 °F) |
MDL Number:
MFCD00011345 CAS Number: 67969-82-8 EINECS: 267-983-8 UN: 3129 ADR: 4.3 (8),II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...