BEAN215570

215570-250ML 215570-1L
BT215570-1LEA 123.45 USD
BT215570-1L BT215570-250ML
BEAN215570
  • RTECS: UF9625000
  • Refractive index: 1.366
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR