Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroethylene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,1,2-Trichloroethene1,1,2-TrichloroethyleneTrichloroethene

215400-1L 215400-4L
BT215400-1LEA 51.47 USD InStock
BT215400-1L BT215400-4L
Trichloroethylene ≥99%
Trichloroethylene
CAS: 79-01-6; EC No: 201-167-4; MDL No: MFCD00000838; RTECS: KX4550000
UN No: UN1710; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: ClCH=CCl2 ; Molecular Formula: C2HCl3; MW: 131.39
Melting Point: -85°; Boiling Point: 87°
Density (g/mL): 1.463; Refractive Index: 1.476
Light Sensitive


Formula: ClCH=CCl₂
MW: 131,39 g/mol
Boiling Pt: 86.7 °C (1013 hPa)
Melting Pt: -73 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Merck Index: 12,09769
https://us.vwr.com/stibo/web/std.lang.all/13/81/741381.jpg
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty