Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromoethanol ≥97%

Supplier: BeanTown Chemical


215370-250G 215370-1KG
BT215370-250GEA 108.22 USD InStock
BT215370-250G BT215370-1KG
2-Bromoethanol ≥97%
2-Bromoethanol
CAS: 540-51-2; EC No: 208-748-1; MDL No: MFCD00002827; RTECS: KJ8225000
UN No: UN2927; Haz Class: 6.1 (8); Packing Group: II
Liquid; Linear Formula: BrCH2CH2OH; Molecular Formula: C2H5BrO; MW: 124.96
Boiling Point: 56-57°/20 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.763; Refractive Index: 1.492
Light Sensitive


Formula: BrH₂CCH₂OH
MW: 124.97 g/mol
Boiling Pt: 149 °C (1013 hPa)
Melting Pt: -80 °C
Density: 1.76 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: 2 - 8°C
MDL Number: MFCD00002827
CAS Number: 540-51-2
EINECS: 208-748-1
UN: 2927
ADR: 6.1,II
Merck Index: 13,03826
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty