Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% ACS

Supplier: BeanTown Chemical

Warning
Synonyms: Tartar emeticPotassium antimony(III) oxide tartrate trihydrateL(+)-Tartaric acid antimony potassium salt trihydratePotassium antimonyl tartrate sesquihydratePotassium Antimony (III) Oxide Tartrate TrihydrateAntimony potassium L(+)-tartrate trihydrateL(+)-Antimony potassium tartrate trihydrate(R,R)-(+)-Tartaric acid antimony potassium salt trihydrate(+)-Antimony potassium L-tartrate trihydratePotassium antimony tartrate trihydrateAntimony potassium tartrate trihydratePotassium antimonyl tartrate trihydrate(R,R)-(+)-Antimony potassium tartrate trihydrateAntimony potassium (+) tartrate trihydrate

215350-500G 215350-25G 215350-100G
BT215350-500GEA 266.92 USD InStock
BT215350-500G BT215350-25G BT215350-100G
Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% ACS
Antimony (III) potassium oxitartrate trihydrate
CAS: 28300-74-5; EC No: 229-436-1; MDL No: MFCD00148863; RTECS: CC6825000
UN No: UN1551; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C8H4K2O12Sb2·3H2O ; MW: 667.87
Melting Point: 300°


Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
MW: 667,88 g/mol
Melting Pt: >300 °C
Density: 2.6 g/cm³ (20 °C)
MDL Number: MFCD00167056
CAS Number: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1,III
Merck Index: 13,00706
Order Now
Specification Test Results
Assay 99.0-103.0%
Loss on drying ≤2.7%
Titrable acid ≤0.020meq/g
Arsenic (As) ≤0.015%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty