Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium ≥99.5% (trace metals basis), ingot

Supplier: BeanTown Chemical


215310-450G
BT215310-450GEA 91.99 USD InStock
BT215310-450G
Sodium ≥99.5% (trace metals basis), ingot
Sodium
CAS: 7440-23-5; EC No: 231-132-9; MDL No: MFCD00085307; RTECS: VY0686000
UN No: UN1428; Haz Class: 4.3; Packing Group: I
Solid, sealed in breakseal ampules; Molecular Formula: Na; MW: 22.99
Melting Point: 97.8°; Boiling Point: 883°; Flash point: 82°C (180°F)
Density (g/mL): 0.968
Air Sensitive


Formula: Na
MW: 22,99 g/mol
Boiling Pt: 881 °C (1013 hPa)
Melting Pt: 97.8 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00085307
CAS Number: 7440-23-5
EINECS: 231-132-9
UN: 1428
ADR: 4.3,I
Merck Index: 13,08641
Order Now
Specification Test Results
Assay 99.5%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty