Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium methoxide ≥98%

Supplier: BeanTown Chemical


215235-100G 215235-500G 215235-25G
BT215235-100GEA 159.93 USD InStock
BT215235-100G BT215235-500G BT215235-25G
Lithium methoxide ≥98%
Lithium methoxide
CAS: 865-34-9; EC No: 212-737-7; MDL No: MFCD00036357
UN No: UN3206; Haz Class: 4.2 (8); Packing Group: II
Powder; Linear Formula: LiOCH3; MW: 37.98
Air Sensitive, Moisture Sensitive


Formula: CH₃LiO
MW: 37,98 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Density: 0.865 g/cm³ (20 °C)
MDL Number: MFCD00036357
CAS Number: 865-34-9
EINECS: 212-737-7
UN: 3206
ADR: 4.2,II
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty