Diammonium tetrachloroplatinate ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Ammonium tetrachloroplatinate(II)
, Ammonium tetrachloroplatinate
215160-5G
215160-1G
BT215160-1GEA
147.47
USD
BT215160-1G
BT215160-5G
Diammonium tetrachloroplatinate ≥99.9% (trace metals basis)
Diammonium tetrachloroplatinate
- RTECS: TP1840000
- Hygroscopic
- Crystalline
Formula:
H₈Cl₄N₂Pt MW: 372.97 g/mol Melting Pt: 140 °C (dec.) Density: 2.936 g/mL |
MDL Number:
MFCD00010885 CAS Number: 13820-41-2 EINECS: 237-499-1 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...