Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium superoxide ≥96%

Supplier: BeanTown Chemical

Synonyms: Potassium dioxide

215050-250G 215050-50G 215050-1KG
BT215050-250GEA 95.89 USD InStock
BT215050-250G BT215050-50G BT215050-1KG
Potassium superoxide ≥96%
Potassium superoxide
CAS: 12030-88-5; EC No: 234-746-5; MDL No: MFCD00082411
UN No: UN2466; Haz Class: 5.1; Packing Group: I
2-10 Micron Powder; Molecular Formula: KO2; MW: 71.10
Melting Point: 400°
Density (g/mL): 2.14


Formula: KO₂
MW: 71,1 g/mol
Melting Pt: 560 °C
Density: 2.14 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00082411
CAS Number: 12030-88-5
EINECS: 234-746-5
UN: 2466
ADR: 5.1,I
Order Now
Specification Test Results
Assay 96%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty