n-Pentane ≥98.0% environmental

Supplier: BeanTown Chemical

Synonyms: Pentane

215020-4L 215020-4X1L 215020-1L
BT215020-4X1LEA 153.71 USD
BT215020-4X1L BT215020-4L BT215020-1L
n-Pentane ≥98.0% environmental
n-Pentane

  • RTECS: RZ9450000
  • Refractive index: 1.358
  • Liquid

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C
Melting Pt: –130 °C
Density: 0.626 g/mL
Flash Pt: –49 °C (–56 °F)
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg

Order Now

Specification Test Results

Assay 98.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR