Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(II) hydride ≥99% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Titanium dihydrideTitanium hydride

214900-50G 214900-5G 214900-250G
BT214900-50G BT214900-5G BT214900-250G
Titanium(II) hydride ≥99% (trace metals basis)
CAS: 7704-98-5; EC No: 231-726-8; MDL No: MFCD00151301; RTECS: XR2130000
UN No: UN1871; Haz Class: 4.1; Packing Group: II
1-3 Micron Powder; Linear Formula: TiH2; MW: 49.92
Melting Point: 450° (decomposes)
Density (g/mL): 3.91
Air Sensitive, Moisture Sensitive


Formula: H₂Ti
MW: 49,9 g/mol
Density: 3.91 g/cm³ (20 °C)
MDL Number: MFCD00151301
CAS Number: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,09473
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty