N,N-Dimethylformamide, anhydrous ≥99.9% amine-free
Supplier: BeanTown Chemical
Synonyms:
NSC 5356
, Formic acid dimethylamide
, Dimethylformamide
, DMFA
, DMF
, Formyldimethylamine
, N-Formyldimethylamine
214860-4L
214860-4X4L
BT214860-4X4LEA
728.97
USD
BT214860-4X4L
BT214860-4L
N,N-Dimethylformamide, anhydrous ≥99.9% amine-free
N,N-Dimethylformamide
- RTECS: LQ2100000
- Refractive index: 1.430
- Liquid
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 °C Melting Pt: –61 °C Density: 0.944 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...